Learn More
Medchemexpress LLC D-Ile-Pro-Arg p-nitroanilide dihydrochloride | 96323-41-0 | MFCD00792772 | >95.0% | 577.50 | C23H38Cl2N8O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1378375MG
Encompass_Preferred
D-Ile-Pro-Arg p-nitroanilide dihydrochloride is a chromogenic peptide substrate used to quantify tissue plasminogen activator (tPA) activity. Enzymatic cleavage releases p-nitroaniline, enabling direct spectrophotometric measurement in biochemical assays. The dihydrochloride salt form improves solubility and handling for routine enzyme kinetics and assay development.
  • Chromogenic substrate for tPA activity assays.
  • Releases p-nitroaniline for spectrophotometric detection.
  • Suitable for enzyme kinetics and biochemical assay development.
  • Provided as a dihydrochloride salt for improved solubility.
  • Available in small packaged quantities for research use.
Catalog No. 50-002-22865