Learn More
Medchemexpress LLC d-KLA Peptide | 286380-05-0 | 99.3% | 1523.99 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP102855MG
Encompass_Preferred
d-KLA Peptide is a synthetic pro-apoptotic peptide that specifically targets mitochondria and induces apoptosis by destroying the mitochondrial membrane. It activates biochemical pathways associated with apoptosis, including the activation of caspase family proteins and PARP (poly ADP ribose polymerase). d-KLA Peptide can also be used to carry and deliver genes or small molecules to enhance anti-tumor effects.
  • Synthetic pro-apoptotic peptide
  • Specifically targets mitochondria
  • Induces apoptosis by destroying the mitochondrial membrane
  • Activates caspase family proteins
  • Activates PARP (poly ADP ribose polymerase)
  • Can be used to carry and deliver genes or small molecules
  • Enhances anti-tumor effects
Catalog No. 50-004-26268