Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-KLVFFA | 342877-55-8 | 99.3% | 723.90 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYP32441MG

Encompass_Preferred

D-KLVFFA is an inhibitor of Amyloid-β assembly, with an IC50 of 2.6 μM, that can be used in Alzheimer's disease study.

  • Inhibitor of Amyloid-β assembly
  • Has an IC50 of 2.6 μM
  • Used in Alzheimer's disease study
  • Appearance is solid, white to off-white
  • Sequence: D-Lys-D-Leu-D-Val-D-Phe-D-Phe-D-Ala
  • Sequence shortening: d-KLVFFA

Catalog No. 50-003-98872


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.