Learn More
Medchemexpress LLC D-kynurenine | 13441-51-5 | MFCD00133224 | 208.21 g/mol | C10H12N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01450250MG
Encompass_Preferred
D-kynurenine is the D-enantiomer of kynurenine, a metabolite of D-tryptophan used as a biochemical research reagent and probe. It functions pharmacologically and analytically, acting at cellular receptors and as a substrate in enzyme assays, and has been reported to influence signaling pathways involved in epithelial-to-mesenchymal transition.
  • Acts as an agonist of GPR109B.
  • Serves as a substrate in fluorometric assays of D-amino acid oxidase.
  • Applicable to studies of the kynurenine metabolic pathway.
  • Reported to modulate epithelial-to-mesenchymal transition via aryl hydrocarbon receptor activation.
Catalog No. 50-003-03258