Learn More
Medchemexpress LLC D-lactate dehydrogenase, microorganism (D-LDH) | 9028-36-8 | 500 U
SDP
Supplier:  Medchemexpress LLC HYP2897500U
Encompass_Preferred
D-Lactate dehydrogenase, Microorganism (D-LDH) is an oxidoreductase that uses NAD+ or NADP+ as an acceptor and acts on the donor CH-OH group. It catalyzes the oxidation of D-lactate to pyruvate. This enzyme is widely found in bacteria and fungi and is frequently used in biochemical research.
  • Catalyzes the oxidation of D-lactate to pyruvate.
  • Uses NAD+ or NADP+ as an acceptor.
  • Isoelectric point: 4.0.
  • Michaelis constant: 1.6x10⁻⁴ M (pyruvate, pH 7.0).
  • Inhibited by Ag+, Hg2+, SH-reagents.
  • Optimum pH: 6.0-7.0.
  • Optimum temperature: 35-40°C.
  • pH stability: 5.0-9.0.
Catalog No. 50-004-00299