Learn More
Medchemexpress LLC [D-Leu-4]-OB3 | 289056-32-2 | 98.9% | 734.82 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP3342100MG
Encompass_Preferred
[D-Leu-4]-OB3 inhibits expressions of pro-inflammatory, proliferative and metastatic genes and PD-L1 expression. It also stimulates expression of pro-apoptotic genes.
  • Inhibits expressions of pro-inflammatory genes.
  • Inhibits expressions of proliferative genes.
  • Inhibits expressions of metastatic genes.
  • Inhibits PD-L1 expression.
  • Stimulates expression of pro-apoptotic genes.
Catalog No. 50-003-99305