Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [D-Leu-4]-OB3 | 289056-32-2 | 98.9% | 734.82 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYP334250MG

Encompass_Preferred

[D-Leu-4]-OB3 is an anti-inflammatory agent that inhibits expressions of pro-inflammatory, proliferative, and metastatic genes, as well as PD-L1 expression. It also stimulates the expression of pro-apoptotic genes.

  • Inhibits expressions of pro-inflammatory, proliferative, and metastatic genes
  • Inhibits PD-L1 expression
  • Stimulates the expression of pro-apoptotic genes
  • Molecular weight: 734.82
  • Formula: C29H50N8O12S
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ser-Cys-Ser-{D-Leu}-Pro-Gln-Thr
  • Sequence shortening: SCS-{D-Leu}-PQT
  • Solubility in DMSO: 50 mg/mL (68.04 mM)

Catalog No. 50-003-97270


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.