Learn More
Medchemexpress LLC D-leucine | 328-38-1 | MFCD00063088 | >98.0% | 131.17 g/mol | C6H13NO2 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0378100G
Encompass_Preferred
D-Leucine is a more potent anti-seizure agent than L-leucine D-leucine potently terminates seizures even after the onset of seizure activity D-leucine but not L-leucine reduces long-term potentiation but had no effect on basal synaptic transmission in vitro[1]
Catalog No. 50-002-69956