Learn More
Medchemexpress LLC D-leucine | 328-38-1 | MFCD00063088 | ≥98.0% | 131.18 | C6H13NO2 | 1mL
SDP
Supplier:  Medchemexpress LLC HYY037810MM/1ML
Encompass_Preferred
D-Leucine is a more potent anti-seizure agent than L-leucine D-leucine potently terminates seizures even after the onset of seizure activity D-leucine but not L-leucine reduces long-term potentiation but had no effect on basal synaptic transmission in vitro[1]
Catalog No. 50-002-68283