Learn More
Medchemexpress LLC D-leucine-d10 | 271247-12-2 | MFCD01321412 | 99.8% | 141.23 | C6H3D10NO2 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0378S25MG
Encompass_Preferred
D-Leucine-d10 is the deuterium labeled D-Leucine D-Leucine is a more potent anti-seizure agent than L-leucine D-leucine potently terminates seizures even after the onset of seizure activity D-leucine but not L-leucine reduces long-term potentiation but had no effect on basal synaptic transmission in vitro[1]
Catalog No. 50-002-68462