Learn More
Medchemexpress LLC D-Luciferin 6 -methy 5mg | 24404-90-8 | 5 MG

Supplier: Medchemexpress LLC HY1157495MG
D-luciferin 6'-methyl ether is a D-luciferin derivative that acts as a potent inhibitor of Photinus pyralis (firefly) luciferase. It is supplied as an off-white to light-yellow solid with high reported purity and is intended for use in luciferase inhibition studies and assay development.
- Potent Photinus pyralis luciferase inhibitor.
- Supplied as an off-white to light-yellow solid.
- Reported purity 99.55%.
- Molecular weight 294.35 g/mol; formula C12H10N2O3S2.
- Storage: 4°C; in solvent store -80°C (6 months) or -20°C (1 month); protect from light.
- Available in small solid quantities and in solution formats (10 mM in DMSO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.