Learn More
Carbosynth LLC. D-Luciferin Firefly, potassium salt, 115144-35-9, MFCD0044928, 1 g
SDP
Supplier:  Carbosynth LLC. L82201G
Encompass_Preferred
Formula: C11H7KN2O3S2, Molecular weight: 318.42 g/mol, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt.
Catalog No. 50-853-139