Learn More
Cayman Chemical D-LucIferInpotasIum salt 1g
SDP
Supplier:  Cayman Chemical 146811 G
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-296-9219