Learn More
Cayman Chemical D-LucIferInsodIum salt 100mg
SDP
Supplier:  Cayman Chemical 14682100 MG
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-297-1596