Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-mannitol-2-13C | 287100-69-0 | MFCD00190335 | 100.0% | 183.16 | C5[13C]H14O6 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0378S310MG

Encompass_Preferred

D-Mannitol-2-13C is the 13C-labeled isotopologue of D-mannitol provided for research use. It is used as an isotopic tracer and NMR reference in biochemical and metabolic studies where a single 13C substitution enables tracking or spectroscopic assignment. Supplied in a 10 mg quantity with high chemical and isotopic purity.

  • 13C-labeled D-mannitol (single label at C-2).
  • High chemical purity (99.95%).
  • Molecular weight 183.16 g/mol.
  • CAS number 287100-69-0.
  • Suitable for NMR, metabolic tracing, and analytical standards.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-95736


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.