Learn More
Medchemexpress LLC D-mannitol-2-13C | 287100-69-0 | MFCD00190335 | 99.0% | 183.16 g/mol | 13C-C6H14O6 | 10 MG

Supplier: Medchemexpress LLC HYN0378S110MG
D-mannitol-13C is the 13C-isotopically labeled form of D-mannitol supplied as a small-format analytical standard for use as a tracer or reference in mass spectrometry and metabolic studies. Isotopic enrichment is typically high, making it suitable for quantitation and tracer experiments.
- 13C-labeled D-mannitol for analytical use.
- Isotopic enrichment: 99.0% (atom% 13C).
- Pack size: 10 MG.
- Molecular formula: C6H14O6 (one 13C).
- Molecular weight: 183.16 g/mol.
- Common applications: tracer, mass spectrometry standard, metabolic studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.