Learn More
Medchemexpress LLC D-manno-2-octulosonic acid, 8-azido-3,8-dideoxy-, ammonium salt | 1380099-68-2 | ≥99.0% | 280.24 | C8H16N4O7 | 10 MG

Supplier: Medchemexpress LLC HY15168510MG
Kdo Azide is an azide-functionalized metabolic labeling reagent for bioorthogonal click chemistry. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) to tag glycoconjugates and other biomolecules for detection, imaging, or conjugation workflows.
- Contains an azide functional group compatible with CuAAC and SPAAC.
- Suitable for metabolic labeling of glycoconjugates and biomolecules.
- High purity: ≥99.0%.
- Solid, white to light yellow appearance.
- Available in multiple package sizes, including milligram quantities.
- Storage: store sealed away from moisture; solids -20°C; in solution -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.