Learn More
AdipoGen D-Mannoheptulose (25 mg)
SDP
Supplier:  AdipoGen AGCR13695M025
Encompass_Preferred
D-Mannoheptulose (25 mg), Adipogen Life Sciences. Chemical. CAS: 3615-44-9. Formula: C7H14O7. MW: 210.2. . Naturally occurring heptose in avocado fruit. Hexokinase (HK) and glucokinase inhibitor in diverse organisms by competing with D-glucose. Useful agent for immunometabolism research. Blocks glucose phosphorylation by blocking hexokinase. Prevents the conversion of glucose to glucose-6-phosphate that can mediate the activation of the carbohydrate response element binding protein and consequently inhibiting the rate limiting step of glycolysis. Blocks glucose oxidation and glucose-mediated insulin release in pancreatic islet cells.
Catalog No. 50-168-7472