Learn More
Medchemexpress LLC D-mannosamine hydrochloride | 5505-63-5 | MFCD00064557 | ≥98.0% | 215.63 | C6H14ClNO5 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW02142550G
Encompass_Preferred
D-Mannosamine hydrochloride is an amino monosaccharide (an amino derivative of D-mannose) supplied as a solid research reagent. It is used in biochemical and cellular studies of carbohydrate metabolism, glycosylation, and cell-surface modification, and is provided with documentation for safe laboratory use.
  • High purity (≥98.0%) suitable for biochemical and cellular assays.
  • Solid, white to off-white form that is easy to handle and weigh.
  • Used in studies of carbohydrate metabolism, glycosylation, and cell-surface modification.
  • Supplied in multiple pack sizes to support small-scale and preparative experiments.
  • Compatible with common aqueous solvents for solution preparation.
Catalog No. 50-003-00131