Learn More
Medchemexpress LLC D-mannosamine hydrochloride | 5505-63-5 | MFCD00064557 | ≥98.0% | 215.63 g·mol⁻¹ | C6H14ClNO5 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW021425100G
Encompass_Preferred
D-Mannosamine hydrochloride is a six-carbon amino sugar used as a biochemical reagent to probe mannose-related pathways and receptors in glycobiology research. It is supplied as a white to off-white solid with high purity suitable for laboratory applications.
  • High purity suitable for analytical and preparative use.
  • White to off-white solid, easy to weigh and dissolve for solution preparation.
  • Useful as a probe for mannose receptor and glycan pathway studies.
  • Stable under refrigerated storage when kept under inert atmosphere.
  • Available in multiple laboratory pack sizes for research flexibility.
Catalog No. 50-003-00489