Learn More
Medchemexpress LLC D-mannosamine hydrochloride | 5505-63-5 | MFCD00064557 | ≥98.0% | 215.63 g/mol | C6H14ClNO5 | 500 MG

Supplier: Medchemexpress LLC HYW021425500MG
D-Mannosamine hydrochloride is the hydrochloride salt of D-mannosamine, a six-carbon amino sugar used as a biochemical reagent in glycobiology and carbohydrate metabolism research. It is supplied as a solid with high purity for laboratory synthesis, cell-based assays, and analytical studies.
- Six-carbon amino sugar useful for glycobiology research.
- Hydrochloride salt form, stable solid for handling and storage.
- High purity suitable for analytical and preparative work.
- Molecular weight 215.63 g/mol.
- Molecular formula C6H14ClNO5.
- Available in multiple pack sizes including 500 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.