Learn More
Medchemexpress LLC D-mannosamine hydrochloride | 5505-63-5 | MFCD09880213 | >98.0% | 215.63 g/mol | C6H14ClNO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02142510G
Encompass_Preferred
D-Mannosamine hydrochloride (CAS 5505-63-5) is a six-carbon amino sugar used as a reagent in biochemical and carbohydrate research. Supplied as the hydrochloride salt, it is characterized by chemical formula C6H14ClNO5 and a molecular weight of 215.63 g/mol, and is provided at research-grade purity for laboratory use.
  • High purity ≥98.0% suitable for analytical and synthetic applications.
  • C hemical formula C6H14ClNO5 and molecular weight 215.63 g/mol.
  • Available in multiple pack sizes including small research quantities and larger lab packs.
  • Store at 4°C under nitrogen; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Used in carbohydrate chemistry, glycosylation studies, and microbiology research.
Catalog No. 50-003-00958