Learn More
Sigma Aldrich Fine Chemicals Biosciences D Mannose powder BioReag500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M6020500G
Encompass
Mannose is an aldohexose carbohydrate. It is isomer of glucose with varying C2 position configuration. Majority of mannose is synthesised from its epimer glucose. It is part of the glycans present in endoplasmic reticulum (ER)/Golgi.
Catalog No. 50-290-7716