Learn More
AdipoGen D Melezitose monohydrate
SDP
Supplier:  AdipoGen CDXM0021G100
Encompass_Preferred
Chemical. CAS 10030-67-8. Formula C18H32O16 . H2O. MW 522.45. Synthetic. Zwitterionic building block for synthesis.
Catalog No. 50-256-0559