Learn More
Medchemexpress LLC D-melibiose | 585-99-9 | 342.30 | C12H22O11 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY107824R25MG
Encompass_Preferred
D-Melibiose (Standard) is an analytical reference standard of the disaccharide D-melibiose, composed of one galactose and one glucose linked α(1→6). It is intended for use as a reference in biochemical assays, metabolomics, and analytical method development. The material is supplied in small quantities for analytical applications and should be stored sealed at -20°C, protected from light and moisture.
  • Composed of one galactose and one glucose linked α(1→6).
  • Intended for analytical and research use in metabolomics and method development.
  • Molecular formula C12H22O11 and molecular weight 342.30 g/mol.
  • Supplied as a small quantity suitable for reference standards and calibration.
  • Store sealed at -20°C, protect from light and moisture.
Catalog No. 50-002-44516