Learn More
Medchemexpress LLC D-Methionine sulfoxide | 21056-56-4 | 98.7% | 100 MG

Supplier: Medchemexpress LLC HY129770100MG
D-methionine sulfoxide is the D-isomer of methionine sulfoxide, an oxidation product of methionine. Methionine is the limiting amino acid in milk or leguminous proteins, easily oxidized during storage or processing. This product is for research use only.
- Solid appearance
- White to off-white color
- Molecular weight of 165.21
- Chemical formula C5H11NO3S
- Classified as others structure
- Initial source is endogenous metabolite
- Solubility in H2O: 125 mg/mL
- In vivo dissolution in PBS: 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.