Learn More
Medchemexpress LLC D-Methionine sulfoxide (hydrochloride) | 1 ML
SDP
Supplier:  Medchemexpress LLC HY129770A10MM/1ML
Encompass_Preferred
D-Methionine sulfoxide hydrochloride is a chemical compound with the formula C5H12CINO3S and a molecular weight of 201.67. It is used as a laboratory chemical and for the manufacture of substances. This D-isomer is provided as a 10mM solution in a 1mL volume.
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications
  • Recommended storage at 4°C, sealed, away from moisture and light
  • Solution storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-40113