Learn More
Abcam D-NMMA monoacetate, L-NMMA control compound, 100MG
Supplier:  Abcam AB144315100MG
Encompass_Preferred
MW 248.28 Da, Purity >98%. Cell-permeable control compound for L-NMMA (ab120137). Inactive, shows no NOS inhibitor activity. Useful tool to investigate non-specific L-NMMA activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5891