Learn More
Cayman Chemical D-OrnIthIn lactam 100mg
SDP
Supplier:  Cayman Chemical 31680100 MG
Encompass_Preferred
A building block; has been used in the synthesis of HCV NS5B inhibitors, as well as αVβ3 integrin receptor antagonists
Catalog No. 50-299-2240