Learn More
Medchemexpress LLC [D-p-Cl-Phe6,Leu17]-VIP TFA | 98.16% | 3456.22 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1159A10MG
Encompass_Preferred
[D-p-Cl-Phe6,Leu17]-VIP TFA is a competitive and selective antagonist of vasoactive intestinal peptide (VIP) receptor, with an IC50 of 125.8 nM. It shows no activity on glucagon, secretin, or GRF receptors.
  • Competitive and selective antagonist of VIP receptor
  • IC50 of 125.8 nM against VIP receptor
  • No activity on glucagon, secretin, or GRF receptors
Catalog No. 50-004-24633