Learn More
Medchemexpress LLC D-Panose | 33401-87-5 | MFCD00151376 | 100.0% | 504.44 | C18H32O16 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1119515MG
Encompass_Preferred
D-Panose is a PAN-type oligosaccharide (trisaccharide) composed of D-glucopyranose units, used as a food ingredient model and as an analytical standard for research applications. It is provided as a high-purity reagent suitable for laboratory assays and characterization.
  • PAN-type oligosaccharide and trisaccharide structure.
  • Suitable as a food ingredient model based on isomaltooligosaccharides (IMOs).
  • High purity for analytical applications (99.95%).
  • Defined molecular formula and weight: C18H32O16, 504.44 g/mol.
  • Available in small quantities for laboratory use.
Catalog No. 50-002-63342