Learn More
Medchemexpress LLC D-(-)-Pantolactone | 599-04-2 | 100.0% | 130.14 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01039650G
Encompass_Preferred
D-(-)-Pantolactone is a crucial intermediate used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It also serves as a calibrant for circular dichroism (CD). Derivatives of D-(-)-Pantolactone have shown good inhibitory activities against FAS and lipid accumulation, suggesting their potential in anti-obesity drug research.
  • Important intermediate for synthesizing active compounds.
  • Functions as a calibrant for circular dichroism (CD).
  • Derivatives show inhibitory activities against FAS and lipid accumulation, relevant for anti-obesity drug research.
Catalog No. 50-004-54024