Learn More
Medchemexpress LLC HY-B0430 100mg , D-Pantothenic acid CAS:79-83-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0430100MG
Encompass_Preferred
HY-B0430 100mg D-Pantothenic acid CAS: 79-83-4 D-Pantothenic acid (Pantothenate) is an essential trace nutrient that functions as the obligate precursor of coenzyme A (CoA). D-Pantothenic acid plays key roles in myriad biological processes, including many that regulate carbohydrate, lipid, protein, and nucleic acid metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1924