Learn More
Medchemexpress LLC D-Pantothenic acid (hemicalcium salt) | 137-08-6 | 238.27 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN068110MM/1ML
Encompass_Preferred
D-Pantothenic acid hemicalcium salt is an orally active vitamin that reduces patulin in apple juice. It removes Reactive Oxygen Species (ROS) and has antibacterial and immunomodulatory activities. It also has the effect of improving osteoporosis and obesity. This compound is suitable for various research applications.
  • Restrains Th17 cell differentiation
  • Inhibits RANKL induced osteoclastogenesis
  • Rescues membrane trafficking defects in human fibroblasts
  • Protects against obesity
  • Alleviates experimental autoimmune encephalomyelitis
  • Alleviates bone loss in osteoporotic models
  • Reduces bacterial growth by regulating immunity
  • Enhances intestinal development and alters microbes
Catalog No. 50-004-16788