Learn More
Medchemexpress LLC D-α-Phenyl-glycine-d5 | 1246817-98-0 | 99.0% | 156.19 | 1 MG

Supplier: Medchemexpress LLC HYW010249S1MG
D-α-Phenyl-glycine-d5 is the deuterium labeled D-α-Phenyl-glycine, intended for research use only. This compound exhibits good stability under recommended storage conditions.
- Formula: C8H4D5NO2
- Purity: 99.0%
- Molecular weight: 156.19
- Solubility: 10 mM in DMSO
- Target: Isotope-labeled compounds
- Appearance: Solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.