Learn More
Medchemexpress LLC D-phenylalanyl-prolyl-arginyl chloromethyl ketone trifluoroacetate | 157379-44-7 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY122542B10MM 1ML
Encompass_Preferred
PPACK TFA is a potent, selective, irreversible thrombin inhibitor supplied as a 10 mM solution in DMSO (1 mL). It is used as an anticoagulant and a biochemical tool to inhibit thrombin activity in blood-sample handling and enzymatic assays.
  • Potent, selective, irreversible thrombin inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity for reproducible experimental results.
  • Molecular weight 564.99 g/mol; CAS 157379-44-7.
  • Store sealed and away from moisture; follow storage instructions for solution and powder.
Catalog No. 50-002-32163