Learn More
Medchemexpress LLC D-phenylglycine methyl ester hydrochloride | 19883-41-1 | MFCD00137487 | 98.0% | 201.65 g/mol | C9H12ClNO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014824100G
Encompass_Preferred
D-phenylglycine methyl ester hydrochloride (CAS 19883-41-1) is the hydrochloride salt of D-phenylglycine methyl ester, used as an amino acid derivative and building block in peptide synthesis and other research applications. The material is supplied as a laboratory-scale reagent suitable for synthesis and analytical work.
  • Hydrochloride form of D-phenylglycine methyl ester.
  • Molecular weight 201.65 g/mol; formula C9H12ClNO2.
  • High purity: 98.0%.
  • Suitable as a chiral building block for peptide synthesis.
  • Stable solid for standard laboratory handling and storage.
Catalog No. 50-003-02596