Learn More
Medchemexpress LLC D-Proline | 344-25-2 | >=98.0% | C5H9NO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY7508710G
Encompass_Preferred
D-Proline, also known as (R)-pyrrolidine-2-carboxylic acid, is a proline isomer known to exhibit high renal and hepatotoxicity in rats. It is utilized in research to study amino acid metabolism and toxicity mechanisms. This product is for research use only and has not been fully validated for medical applications.
  • Proline isomer.
  • Exhibits high renal and hepatotoxicity in rats.
  • Suitable for studying amino acid metabolism and toxicity mechanisms.
  • Appears as a white to off-white solid.
  • For research use only.
Catalog No. 50-004-34369