Learn More
Alkali Scientific D (+) Raffinose Pentahydrate, 5 Kg
SDP
Supplier:  Alkali Scientific DRA005CP
Encompass
D (+) Raffinose Pentahydrate (5 Kg) is a premium-grade carbohydrate reagent used in biochemical and laboratory applications. Raffinose, a trisaccharide composed of galactose, glucose, and fructose, is commonly utilized in studies involving plant physiology, microbial fermentation, and metabolic research. The 5 Kg quantity is suitable for large-scale studies, offering ample supply for high-volume experiments. This substance is used for its ability to provide a stable environment for enzymes and other biological systems. The product is supplied by Alkali Scientific in high purity, ensuring consistency in experiments and analysis.
Catalog No. 50-006-60162