Learn More
Medchemexpress LLC D-Ribonolactone | 5336-08-3 | ≥98.0% | C₅H₈O₅ | 5 G
SDP
Supplier:  Medchemexpress LLC HY766915G
Encompass_Preferred
D-Ribonolactone is a sugar lactone and an inhibitor of β-galactosidase of *Escherichia coli*, with a Ki of 26 mM. It appears as a white to off-white solid and has a molecular weight of 148.11. This compound is classified as a saccharide, monosaccharide, and an endogenous metabolite, intended for research use only.
  • Inhibits β-galactosidase (Ki: 26 mM)
  • Presented as a white to off-white solid
  • Molecular weight is 148.11
  • Classified as a saccharide and monosaccharide
  • Functions as an endogenous metabolite
  • Suitable for research applications only
Catalog No. 50-004-33726