Learn More
Medchemexpress LLC D-ribose-13C-4 | 99.0% | 151.12 g/mol | C4(13C)H10O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW018772S51MG
Encompass_Preferred
D-Ribose-13C-4 is a stable-isotope labeled D-ribose with the 13C label at the C-4 position, supplied as a high-purity tracer and internal standard for quantitative NMR, GC-MS, and LC-MS and for metabolic studies involving ATP and protein glycation.
  • High isotopic incorporation at the C-4 position.
  • Purity: 99.0%.
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High solubility in water and DMSO (≈100 mg/mL).
  • Stable under recommended storage conditions for powder and solutions.
Catalog No. 50-003-02034