Learn More
Medchemexpress LLC D-ribose-d2 | 478506-32-0 | 98.0% | 152.14 g·mol⁻1 | C5H8D2O5 | 1 MG

Supplier: Medchemexpress LLC HYW018772S101MG
Deuterium-labeled D-ribose is a stable isotope reagent used as an analytical standard and tracer in metabolic and biochemical research. Supplied as a powder with defined storage instructions, it supports studies of ATP metabolism, protein glycation, and metabolic pathways while providing reliable isotopic labeling for analytical workflows.
- Deuterium-labeled sugar for isotope tracing.
- High purity: 98.0%.
- Molecular formula C5H8D2O5; molecular weight 152.14 g·mol⁻1.
- Available in multiple pack sizes, including 1 mg.
- Powder storage: -20°C (stable up to 3 years); in solvent: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.