Learn More
Medchemexpress LLC D-ribose-d6 | 99.9% | 156.17 g/mol | C5H4D6O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW018772S121MG
Encompass_Preferred
D-Ribose-d6 is a deuterium-labeled form of D-ribose used as an isotopic standard and tracer in analytical and biochemical research. It provides a mass-shifted reference for accurate identification and quantitation in MS and NMR studies, and is suitable for metabolic labeling and tracer experiments requiring deuterium enrichment.
  • Deuterium-labeled (d6) isotopic standard.
  • High purity, suitable for analytical use.
  • Molecular weight 156.17 g/mol; formula C5H4D6O5.
  • Compatible with mass spectrometry and NMR applications.
  • Packaged for small-scale laboratory research.
Catalog No. 50-003-00034