Learn More
Medchemexpress LLC D-saccharic acid 1,4-lactone hydrate | 61278-30-6 | MFCD00149334 | 98.0% | 210.14 g/mol | C6H10O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY134453A5MG
Encompass_Preferred
D-saccharic acid 1,4-lactone hydrate is a crystalline β-glucuronidase inhibitor supplied as a high-purity solid for biochemical research. It is used in enzyme inhibition studies, buffer formulations, and other laboratory assays where inhibition of β-glucuronidase or study of glucaric-acid derivatives is required.
  • High purity (98.0%).
  • Solid form, white to off-white appearance.
  • Molecular formula C6H10O8; molecular weight 210.14 g/mol.
  • Available in small milligram quantities and as a DMSO solution.
  • Suitable for enzyme inhibition studies and biochemical assays.
Catalog No. 50-002-22022