Learn More
Lonza Walkersville D-SAEC-CF - Cystic Fibrosis
Supplier:  Lonza Walkersville CC2933
Encompass
Cystic fibrosis is a disease that causes thick sticky mucus to build up in the lungs digestive tract and other areas of the body It is one of the most common chronic lung diseases in children and young adults Lonza s Human Small Airway Epithelial Cells (SAEC) are isolated from cystic fibrosis human lung tissue Cryopreserved Clonetics SAECs are guaranteed through 15 population doublings and stain positive for cytokeratin 19 All cells testnegative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B andhepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96365