Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp D-SALICIN, 100 GR
SDP

Supplier:  Research Products International Corp S23200100.0

Encompass_Preferred

CAS Number: 138-52-3
Molecular Formula: C13H18O7
Molecular Weight: 286.3
Chemical Purity: >99%
Soluble in: Water
Store at: Room Temperature

Catalog No. 50-489-024


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.