Learn More
Medchemexpress LLC D-serine | 312-84-5 | MFCD00004269 | 98.0% | 105.09 g/mol | C3H7NO3 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10080825G
Encompass_Preferred
D-serine is the D-enantiomer of the amino acid serine and functions as a co-agonist at NMDA-type glutamate receptors. Provided as a research-grade chemical for analytical and laboratory use, it is commonly employed in neuropharmacology, studies of synaptic plasticity, and biochemical assays. The reagent is supplied in a 25 g package and is documented with a reported purity of 98%.
  • High purity suitable for research applications.
  • Acts as an NMDA receptor co-agonist relevant to neurobiological studies.
  • Provided in a 25 g package for laboratory-scale experiments.
  • Molecular weight 105.09 g/mol for accurate solution preparation.
  • Datasheet and certificate of analysis available for quality verification.
Catalog No. 50-002-12176