Learn More
Abcam D-Serine (mM/ml), NMDA receptor co-agonist, 100MM_ML
Supplier:  Abcam AB146671100MM_ML
Encompass_Preferred
MW 105.09 Da, Purity >98%. Agonist at the NMDA glycine binding site and the inhibitory post-synaptic glycine receptor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4533