Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-serine, phenylmethyl ester, hydrochloride | 151651-44-4 | MFCD00191017 | 99.4% | 231.68 | C10H14ClNO3 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW03588610G

Encompass_Preferred

Benzyl D-serinate hydrochloride is the benzyl ester hydrochloride salt of D-serine, supplied as a white to off-white solid for use in biochemical and medicinal chemistry research. It serves as an amino acid derivative and synthetic building block in studies involving serine analogs and receptor modulation.

  • White to off-white solid.
  • Molecular formula C10H14ClNO3.
  • Molecular weight 231.68 g/mol.
  • Purity approximately 99.4%.
  • Used as an amino acid derivative and synthetic building block.
  • Available in small research pack sizes.

Catalog No. 50-002-99403


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.