Learn More
Medchemexpress LLC D-serine (R-serine) | 312-84-5 | MFCD00004269 | 98.0% | 105.09 g/mol | C3H7NO3 | 500 MG

Supplier: Medchemexpress LLC HY100808500MG
D-Serine (R-serine) is an endogenous amino acid that functions as a co-agonist at NMDA-type glutamate receptors and is used as a research reagent to study glutamatergic signaling, synaptic plasticity, and neurobiological processes. Supplied as a powder for laboratory use.
- High purity: 98.0%.
- Research reagent for NMDA receptor and synaptic studies.
- Stable powder with recommended storage at -20°C for long-term.
- Small-molecule formula C3H7NO3; molecular weight 105.09 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.